missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL12/SDF-1 alpha Quantikine ELISA Kit

Product Code. 18702384
Change view
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702384 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702384 Supplier R&D Systems™ Supplier No. MCX120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse CXCL12/SDF-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL12/SDF-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CXCL12/SDF-1 alpha in cell culture supernates (10 ul), serum (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 - 10ng/mL
Assay Sensitivity 0.069ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse SDF-1 alpha
For Use With (Application) ELISA
Gene ID (Entrez) 20315
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (10uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF-1, SDF1 alpha, SDF1a, SDF1B, TLSF, TPAR1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.