missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CXCL10/IP-10/CRG-2 DuoSet ELISA

Product Code. p-4754718
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
This item is not returnable. View return policy

Product Code. 30021334

Brand: R&D Systems™ DY466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse CXCL10/IP-10/CRG-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL10/IP-10/CRG-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CXCL10/IP-10/CRG-2.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Kit
Quantity 1 Kit
Species Mouse
Conjugate Biotin
Assay Range 62.50 to 4000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 15 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 15945
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.