missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL10/IP-10/CRG-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB466-SP
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL10/IP-10/CRG-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3627 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 134013 | |
| Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P17515 | |
| Cxcl10 | |
| E. coli-derived recombinant mouse CXCL10/IP-10/CRG-2 Ile22-Pro98 Accession # P17515 | |
| 25 μg | |
| Primary | |
| Detects mouse CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) CXCL1, 2, 3, 5, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , 13, rmCXCL1, 2, 6, 9, 11, 12/SDF-1 alpha , 13, rpCXCL8, rrCXCL1, 3/CINC-2 alpha , 3/CINC-2 beta | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction