missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CXCL1 (GRO alpha) Recombinant Protein, PeproTech®

Product Code. 17841913
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17841913

Brand: Gibco™ 25011250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

250-11-1MG will be provided as 2 x 500 μg (250-11-500UG). Recombinant Murine KC is a 7.8 kDa protein consisting of 72 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12850
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 14825
Molecular Weight (g/mol) 7.8 kDa
Name Mouse CXCL1 (GRO alpha)
Purification Method Purified
Quantity 250 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO α; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROα; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51
Common Name CXCL1 (GRO alpha)
Gene Symbol Cxcl1
Biological Activity Determined by its ability to chemoattract total human neutrophil population using a concentration range of 10.0-100.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence APIANELRCQ CLQTMAGIHL KNIQSLKVLP SGPHCTQTEV IATLKNGREA CLDPEAPLVQ KIVQKMLKGV PK
Content And Storage -20°C
Activity Determined by its ability to chemoattract total human neutrophil population using a concentration range of 10.0-100.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.