missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXADR Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2654
This item is not returnable.
View return policy
Description
CXADR Polyclonal specifically detects CXADR in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| CXADR | |
| Polyclonal | |
| Unconjugated | |
| P97792 | |
| CXADR | |
| Mouse myeloma cell line NS0-derived recombinant mouse CXADR Leu20-Gly237 Accession # P97792 | |
| 100 μg | |
| Primary | |
| Detects mouse CXADR in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human CXADR is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 12.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3 BP, CVB3-binding protein, hCAR, HCVADR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1525 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction