missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CTACK (CCL27) Recombinant Protein, PeproTech®

Product Code. 17819982
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17819982

Brand: Gibco™ 25026250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine CTACK is a 10.9 kDa protein containing 95 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL27 gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The protein encoded by this gene is chemotactic for skin-associated memory T lymphocytes. This cytokine may also play a role in mediating homing of lymphocytes to cutaneous sites. It specifically binds to chemokine receptor 10 (CCR10). Studies of a similar murine protein indicate that these protein-receptor interactions have a pivotal role in T cell-mediated skin inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z1X0
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 20301
Molecular Weight (g/mol) 10.9 kDa
Name Mouse CTACK (CCL27)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias ALP; AW558992; CC chemokine ILC; C-C motif chemokine 27; C-C motif chemokine ligand 27; Ccl27; Ccl27a; chemokine (C-C motif) ligand 27; chemokine (C-C motif) ligand 27 A; CTACK; CTAK; cutaneous T-cell attracting chemokine; cutaneous T-cell-attracting chemokine; ESkine; IL-11 R-alpha-locus chemokine; IL-11 Ralpha-locus chemokine; ILC; PESKY; SCYA27; Scya27a; Skinkine; skinskine; small inducible cytokine A27; small inducible cytokine A27a; small inducible cytokine subfamily A (Cys-Cys), member 27; small-inducible cytokine A27
Common Name CCL27
Gene Symbol Ccl27a
Biological Activity Determined by its ability to chemoattract human peripheral blood lymphocytes using a concentration range of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPLPSSTSCC TQLYRQPLPS RLLRRIVHME LQEADGDCHL QAVVLHLARR SVCVHPQNRS LARWLERQGK RLQGTVPSLN LVLQKKMYSN PQQQN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.