missing translation for 'onlineSavingsMsg'
Learn More

Mouse Crossveinless-2/CV-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication

Brand:  R&D Systems MAB2299

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18142121

  • £333.68 / 100µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Crossveinless-2/CV-2/BMPER Monoclonal specifically detects Crossveinless-2/CV-2/BMPER in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Crossveinless-2/CV-2/BMPER
Monoclonal
LYOPH
Western Blot 1 ug/mL
AAH66153
BMPER
Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2 Val34-Arg685 Accession # AAH66153
100 μg
Primary
Detects mouse Crossveinless-2/CV-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human CV-2 is observed.
Mouse
Purified
Western Blot
349920
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BMP binding endothelial regulator, BMP-binding endothelial regulator precursor protein, BMP-binding endothelial regulator protein, BMPER, Bone morphogenetic protein-binding endothelial cell precursor-derived regulator, CRIM3, crossveinless 2, crossveinless-2, Cv2, CV-2, hCV2, KIAA1965, Protein crossveinless-2
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
168667
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only