missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Crossveinless-2/CV-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF2299-SP
This item is not returnable.
View return policy
Description
Crossveinless-2/CV-2/BMPER Polyclonal specifically detects Crossveinless-2/CV-2/BMPER in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Crossveinless-2/CV-2/BMPER | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP binding endothelial regulator, BMP-binding endothelial regulator precursor protein, BMP-binding endothelial regulator protein, BMPER, Bone morphogenetic protein-binding endothelial cell precursor-derived regulator, CRIM3, crossveinless 2, crossveinless-2, Cv2, CV-2, hCV2, KIAA1965, Protein crossveinless-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 168667 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| AAH66153 | |
| BMPER | |
| Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2 (R&D Systems, Catalog # 2299-CV) Ala39-Arg685 Accession # AAH66153 | |
| 25 μg | |
| Primary | |
| Detects mouse Crossveinless-2/CV-2 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human CV-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction