missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CRISP-4 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5017
This item is not returnable.
View return policy
Description
CRISP-4 Polyclonal specifically detects CRISP-4 in Mouse samples. It is validated for Western Blot.
Specifications
| CRISP-4 | |
| Polyclonal | |
| Unconjugated | |
| NP_084309 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10321 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| Crisp4 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CRISP-4 Leu23-Lys250 Accession # NP_084309 | |
| 100 μg | |
| Primary | |
| Detects mouse CRISP-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse CRISP-1, -2, and -3 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction