missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CREG Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF1697
Additional Details : Weight : 0.00970kg
Description
CREG Polyclonal specifically detects CREG in Mouse samples. It is validated for Western Blot.Specifications
CREG | |
Polyclonal | |
Unconjugated | |
NP_035934 | |
CREG1 | |
E. coli-derived recombinant mouse CREG Arg32-Gln220 Accession # NP_035934 | |
100 μg | |
Primary | |
Detects mouse CREG in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human CREG is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, CREG1, protein CREG1 | |
Goat | |
Affinity Purified | |
RUO | |
8804 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only