missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Contactin-5 (NP_001164258.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | Contactin-5 |
| Accession Number | NP_001164258.1 |
| Gene ID (Entrez) | 53942 |
| Gene Alias | CNTN5, contactin 5, contactin-5, Contactin5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction