missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Complement Component C5a Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2150-SP
This item is not returnable.
View return policy
Description
Complement Component C5a Monoclonal specifically detects Complement Component C5a in Mouse samples. It is validated for Western Blot.
Specifications
| Complement Component C5a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P06684 | |
| Hc | |
| E. coli-derived recombinant mouse C5a Asn679-Arg755 Accession # P06684 | |
| 25 μg | |
| Primary | |
| Detects mouse Complement Component C5a in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) C3a, rmC3d, or rhC5a. | |
| Mouse | |
| Purified |
| Western Blot | |
| 295103 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C5, C5A, HE | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 727 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction