missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Complement C5b-9 (soluble) ELISA Kit

Product Code. 30245133
Click to view available options
:
96 Tests
This item is not returnable. View return policy

Product Code. 30245133

Brand: Invitrogen™ EEL207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The complement system is a fundamental part of innate immunity, consisting of over 45 genes that encode various complement components, regulators, and receptors. The complement C5b-9 complex, also known as the Membrane Attack Complex (MAC), plays a crucial role in the immune defense mechanism. It includes the sequential assembly of proteins C5b, C6, C7, C8, and multiple copies of C9. This complex forms transmembrane channels that puncture the cell membrane of target cells, leading to cell lysis. Specifically, MAC has significant implications in mediating cell death in both pathogenic organisms and cancer cells. The genes involved are clustered across the human genome, mainly organized in three significant loci. The understanding of the C5b-9 complex's function extends beyond its antimicrobial roles to include its interactions with tumor cells, where it can induce cell death and influence cancer progression.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06683
Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format ELISA Microplate (Dismountable)
Product Type ELISA
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 109828, 12274, 12279
Gene Symbol C6, C7, C9
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Complement C5b-9
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.