missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse COCO Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3356
This item is not returnable.
View return policy
Description
COCO/DAND5 Polyclonal specifically detects COCO/DAND5 in Mouse samples. It is validated for Western Blot.
Specifications
| COCO/DAND5 | |
| Polyclonal | |
| Unconjugated | |
| NP_957679 | |
| DAND5 | |
| E. coli- and mouse myeloma cell line NS0-derived recombinant mouse COCO Arg24-Leu185 Accession # NP_957679 | |
| 100 μg | |
| Primary | |
| Detects mouse COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human COCO is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 199699 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction