missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Coagulation Factor VII Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB33051-SP
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Coagulation Factor VII | |
| Monoclonal | |
| LYOPH | |
| Western Blot, Immunohistochemistry 8-25 ug/mL | |
| P70375 | |
| F7 | |
| Chinese hamster ovary cell line CHO-derived recombinant human mouse Coagulation Factor VII Ala42-Leu446 Accession # P70375 | |
| 25 μg | |
| Primary | |
| Detects mouse Coagulation Factor VII in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Factor X, rhFactor XI, rhPROC, rhThrombin, rhKallikrein 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, or Kallikrein B1 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 406707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, F7, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2155 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction