missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Coagulation Factor III/Tissue Factor Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Marke: R&D Systems BAF3178
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Coagulation Factor III/Tissue Factor Polyclonal specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blot.
Spezifikation
| Coagulation Factor III/Tissue Factor | |
| Polyclonal | |
| Biotin | |
| P20352 | |
| F3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Coagulation Factor III/Tissue Factor Ala29-Glu251 Accession # P20352 | |
| 50 μg | |
| Primary | |
| Detects mouse Coagulation Factor III/Tissue Factor in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human TF is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2152 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur