missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CMG-2/ANTXR2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3636-SP
This item is not returnable.
View return policy
Description
CMG-2/ANTXR2 Polyclonal specifically detects CMG-2/ANTXR2 in Mouse samples. It is validated for Western Blot.
Specifications
| CMG-2/ANTXR2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| anthrax toxin receptor 2, ANTXR2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, cI-35, CMG2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF, JHS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 118429 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6DFX2 | |
| ANTXR2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CMG-2/ANTXR2 Gly26-Gly318 Accession # Q6DFX2 | |
| 25 μg | |
| Primary | |
| Detects mouse CMG-2/ANTXR2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human CMG-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction