missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CL-P1/COLEC12 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB31301
This item is not returnable.
View return policy
Description
CL-P1/COLEC12 Monoclonal specifically detects CL-P1/COLEC12 in Mouse samples. It is validated for Western Blot.
Specifications
| CL-P1/COLEC12 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8K4Q8 | |
| Colec12 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CL-P1/COLEC12 Ala101-Leu742 Accession # Q8K4Q8 | |
| 100 μg | |
| Primary | |
| Detects mouse CL-P1/COLEC12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human CL-P1 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 350601 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CL-P1, CLP1hCL-P1, COLEC12, collectin placenta 1, Collectin placenta protein 1, collectin sub-family member 12, collectin-12, NSR2, Nurse cell scavenger receptor 2, SCARA4, SCARA4NSR2, Scavenger receptor class A member 4, scavenger receptor class A, member 4, SRCL Type I, SRCLScavenger receptor with C-type lectin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 81035 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction