missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Chordin ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Mouse Chordin quantitates mouse Chordin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Chordin.
This gene encodes a secreted protein that dorsalizes early vertebrate embryonic tissues by binding to ventralizing TGF-beta-like bone morphogenetic proteins and sequestering them in latent complexes. The encoded protein may also have roles in organogenesis and during adulthood. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequence has not been determined.
Specifications
Specifications
| Accession Number | Chd; Chordin; Chrd; UNQ217/PRO243 |
| Assay Range | 49 to 12,000 pg/mL |
| Assay Sensitivity | 49 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 12667 |
| Gene Symbol | CHRD |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction