missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CHL-1/L1CAM-2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2147
This item is not returnable.
View return policy
Description
CHL-1/L1CAM-2 Polyclonal specifically detects CHL-1/L1CAM-2 in Mouse samples. It is validated for Western Blot.
Specifications
| CHL-1/L1CAM-2 | |
| Polyclonal | |
| Biotin | |
| CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10752 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CHL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CHL-1/L1CAM-2 | |
| 50 μg | |
| Primary | |
| Detects mouse CHL-1/L1CAM-2 in Western blots. In Western blots, approximately 75% cross-reactivity with recombinant human CHL-1. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction