missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CHL-1/L1CAM-2 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
£336.92
Specifications
Antigen | CHL-1/L1CAM-2 |
---|---|
Clone | 316316 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
CHL-1/L1CAM-2 Monoclonal specifically detects CHL-1/L1CAM-2 in Mouse samples. It is validated for Western Blot.Specifications
CHL-1/L1CAM-2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
BAC30699 | |
10752 | |
Mouse myeloma cell line NS0-derived recombinant mouse CHL-1/L1CAM-2 Ala25-Gln1043 (Leu227-Gln242 del & Ala243Ser) Accession # BAC30699 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CHL-1/L1CAM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CHL-1 is observed. |
316316 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CALL, CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), CHL1, FLJ44930, L1CAM-2, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
CHL1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only