missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Chitotriosidase/CHIT1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5325-SP
This item is not returnable.
View return policy
Description
Chitotriosidase/CHIT1 Polyclonal specifically detects Chitotriosidase/CHIT1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Chitotriosidase/CHIT1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHI3, CHIT1, CHITCHI3, chitinase 1 (chitotriosidase), Chitinase-1, chitotriosidase-1, EC 3.2.1.14, FLJ00314, MGC125322, plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1118 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9D7Q1 | |
| CHIT1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Chitotriosidase/CHIT1 Ala22-Ser464 Accession # Q9D7Q1 | |
| 25 μg | |
| Primary | |
| Detects mouse Chitotriosidase/CHIT1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human CHIT1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction