missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ChemR23 PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB7610P-100UG
This item is not returnable.
View return policy
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| ChemR23/CMKLR1 | |
| Monoclonal | |
| PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1240 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 477806 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, cmklr1, Dez, Gpcr27, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
| NS0 mouse myeloma cell line transfected with mouse ChemR23 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction