missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cerberus 1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1986
Additional Details : Weight : 0.00970kg
Description
Cerberus 1 Polyclonal specifically detects Cerberus 1 in Mouse samples. It is validated for Western Blot.Specifications
Cerberus 1 | |
Polyclonal | |
Unconjugated | |
O55233 | |
CER1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cerberus 1 Asp41-Pro272 Accession # O55233 | |
100 μg | |
Primary | |
Detects mouse Cerberus 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Cerberus is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CER1, cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
Goat | |
Affinity Purified | |
RUO | |
9350 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only