missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CDCP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£167.00 - £501.00
Specifications
| Antigen | CDCP1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789017
![]() |
R&D Systems
AF4515-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18092012
![]() |
R&D Systems
AF4515 |
100 μg |
£501.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDCP1 Polyclonal specifically detects CDCP1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CDCP1 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Sheep | |
| Mouse | |
| CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
| CDCP1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q5U462 | |
| 64866 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CDCP1 Arg25-Ala667 Accession # Q5U462 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse CDCP1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human CDCP1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title