missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD72 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1279-SP
This item is not returnable.
View return policy
Description
CD72 Polyclonal specifically detects CD72 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| CD72 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD72, CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Ly-19.2, Ly-32.2, Lyb-2, LYB2lyb-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 971 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P21855 | |
| CD72 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD72 Arg117-Asn354 Accession # P21855 | |
| 25 μg | |
| Primary | |
| Detects mouse CD72 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human CD72 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction