missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD68/SR-D1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10114-100
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1 | |
| Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession No. P31996 | |
| Primary | |
| Detects mouse CD68 in direct ELISAs. | |
| Mouse | |
| IgG |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 2449C | |
| Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 3-25 μ/mL, CyTOF-ready | |
| P31996 | |
| Rabbit | |
| 100 μg | |
| 968 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction