missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD5L Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2834-SP
This item is not returnable.
View return policy
Description
CD5L Polyclonal specifically detects CD5L in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CD5L | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CD5L, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 922 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9QWK4 | |
| CD5L | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD5L Glu22-Val352 Accession # Q9QWK4 | |
| 25 μg | |
| Primary | |
| Detects mouse CD5L in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD5L is observed and less than 1% cross-reactivity with recombinant mouse CD5 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction