missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD48/SLAMF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3327
This item is not returnable.
View return policy
Description
CD48/SLAMF2 Polyclonal specifically detects CD48/SLAMF2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| CD48/SLAMF2 | |
| Polyclonal | |
| Unconjugated | |
| BAE96326 | |
| CD48 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD48/SLAMF2 Phe23-Arg216 Accession # BAE96326 | |
| 100 μg | |
| Primary | |
| Detects mouse CD48/SLAMF2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human CD48, recombinant mouse (rm) OX40, rm2B4, and rmNTB-A, and rmCD2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| BCM1, BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 962 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction