missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD40/TNFRSF5 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB4401V-100UG
This item is not returnable.
View return policy
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD40/TNFRSF5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P27512 | |
| CD40 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD40/TNFRSF5 Val24-Arg193 Accession # P27512 | |
| RUO | |
| 958 | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 65924 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse CD40/TNFRSF5 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant human CD40/TNFRSF5, recombinant mouse (rm) CD40 Ligand, rmCD27/TNFRSF7, rmCD30/TNFRSF8, or rmFas/TNFRSF6 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction