missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD39L3/ENTPD3 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF4464
This item is not returnable.
View return policy
Description
CD39L3/ENTPD3 Polyclonal specifically detects CD39L3/ENTPD3 in Mouse samples. It is validated for Western Blot.
Specifications
| CD39L3/ENTPD3 | |
| Polyclonal | |
| Biotin | |
| AAH79871 | |
| ENTPD3 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD39L3/ENTPD3 Gln44-Pro485 Accession # AAH79871 | |
| 50 μg | |
| Primary | |
| Detects mouse CD39L3/ENTPD3 in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human CD39L3 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| ATPDase, CD39 antigen-like 3, CD39L3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, ENTPD3, FLJ93839, HB6, HB6NTPDase 3, NTPDase-3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 956 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction