missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD39L2/ENTPD6 Antibody, Novus Biologicals™

Rat Monoclonal Antibody
£169.00 - £522.00
Specifications
| Antigen | CD39L2/ENTPD6 |
|---|---|
| Clone | 774308 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766225
![]() |
R&D Systems
MAB6288-100 |
100 μg |
£522.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756095
![]() |
R&D Systems
MAB6288-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD39L2/ENTPD6 Monoclonal antibody specifically detects CD39L2/ENTPD6 in Mouse samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| CD39L2/ENTPD6 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD39 antigen-like 2, CD39L2, CD39L2NTPDase-6, CD39-like 2, dJ738P15.3, DKFZp781G2277, DKFZp781K21102, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 6, ectonucleoside triphosphate diphosphohydrolase 6 (putative function), ectonucleoside triphosphate diphosphohydrolase 6 (putative), ENTPD6, FLJ36711, IL-6SAG, IL6ST2, interleukin 6 signal transducer-2, NTPD3, NTPDase 6 | |
| Chinese Hamster Ovary cell line, CHO-derived mouse CD39L2/ENTPD6, Lys33-Leu455, Accession # 742115 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 774308 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 742115 | |
| 955 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title