missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD36/SR-B3 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £370.00
Specifications
| Antigen | CD36/SR-B3 |
|---|---|
| Clone | 324216 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722999
![]() |
R&D Systems
MAB2519-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132641
|
R&D Systems
MAB2519 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for Western Blot.Specifications
| CD36/SR-B3 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q08857 | |
| 948 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD36/SR-B3 Gly30-Lys439 Accession # Q08857 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD36/SR-B3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD36 is observed. |
| 324216 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, Collagen R, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, SRB3, SR-B3, Thrombospondin R, Thrombospondin receptor | |
| CD36 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title