missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD31/PECAM-1 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB3628
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q08481 | |
| PECAM1 | |
| NS0 mouse myeloma cell line transfected with mouse CD31/PECAM-1 Glu18-Lys590 Accession # Q08481 | |
| 100 μg | |
| Primary | |
| Detects mouse CD31/PECAM-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) DCC, rmICAM-1, -2, -5, rmMAdCAM-1, rmVCAM-1, recombinant porcine CD31/PECAM-1, recombinant human (rh) CD31/PECAM-1, rhCEACAM-1, rhSIGIRR, rhICAM-3, or rhICAM-4 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 693102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5175 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction