missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD300b/LMIR5 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2580
This item is not returnable.
View return policy
Description
CD300b/LMIR5/CD300LB Polyclonal specifically detects CD300b/LMIR5/CD300LB in Mouse samples. It is validated for Western Blot.
Specifications
| CD300b/LMIR5/CD300LB | |
| Polyclonal | |
| Unconjugated | |
| Q3U497 | |
| CD300LB | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD300b/LMIR5 Ile18-Tyr157 Accession # Q3U497 | |
| 100 μg | |
| Primary | |
| Detects mouse CD300b/LMIR5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human LMIR5 is observed and less than 5% cross-reactivity with recombinant human (rh) LMIR1, rhLMIR2, recombinant mouse LMIR3, and rhLMIR6 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CD300LB, CLM7, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Gm253, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, Triggering receptor expressed on myeloid cells 5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 124599 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction