missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD30/TNFRSF8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF852-SP
This item is not returnable.
View return policy
Description
CD30/TNFRSF8 Polyclonal specifically detects CD30/TNFRSF8 in Mouse samples. It is validated for Western Blot, Agonist Activity, Immunocytochemistry.
Specifications
| CD30/TNFRSF8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects CD30/TNFRSF8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD30 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Agonist Activity, Immunocytochemistry 5-15 ug/mL | |
| Q60846 | |
| TNFRSF8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD30/TNFRSF8 Phe19-Thr281 Accession # Q60846 | |
| 25 μg | |
| Agonist Activity | |
| 943 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction