missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD3 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB4841S-025
This item is not returnable.
View return policy
Description
CD3 Monoclonal specifically detects CD3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cd247 | |
| T cell hybridoma D1 | |
| 25 Tests | |
| Primary | |
| Reacts with mouse TCR-associated CD3 complex that occurs on thymocytes and mature T cells. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 17A2 | |
| Flow Cytometry 5 uL/10^6 cells | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 916 | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction