missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD28 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB4831-SP
This item is not returnable.
View return policy
Description
CD28 Monoclonal specifically detects CD28 in Mouse samples. It is validated for Western Blot.
Specifications
| CD28 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 940 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2c |
| Western Blot | |
| 61109 | |
| Western Blot 1 ug/mL | |
| P31041 | |
| CD28 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CD28 Lys21-Lys149 Accession # P31041 | |
| 25 μg | |
| Primary | |
| Detects mouse CD28 in direct ELISAs and Western blots. Does not cross-react with recombinant human CD28, recombinant mouse (rm) CTLA-4, rmICOS, or rmPD-1. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction