missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD27/TNFRSF7 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5741U-100UG
This item is not returnable.
View return policy
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD27/TNFRSF7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P41272 | |
| CD27 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD27/TNFRSF7 Thr21-Arg182 Accession # P41272 | |
| RUO | |
| 939 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 137915 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD27, CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse CD27 in direct ELISAs and Western blots. In Western blots; does not cross-react with recombinant human (rh) CD27, recombinant mouse (rm) CD40, rmGITR, rhDR6, rhTRAIL R4, rmHVEM, rmCD30, rmRANK, rmFas, rmTRAIL R2, rhTRADD, rm4-1BB, rmLT beta R, rmOPG, rmNGFR, rmDR3, rmBAFF R, rhTRAIL R3, rmTAJ, rmEDAR, rmOX-40, rmTWEAK R, rmTNF R1, rhRELT, or rmTRAIL R1. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction