missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CD200 (soluble) Fc Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Description
CD200 is a type-1 membrane glycoprotein that contains two immunoglobulin domains, and belongs to the immunoglobulin superfamily. CD200 is a broadly distributed cell surface glycoprotein that interacts with a receptor on myeloid cells (CD200R) and is implicated in locally preventing macrophage activation. Distant, but recognizable, homologues of CD200 have been identified in many herpesviruses and poxviruses. Studies of the related genes in mouse and rat suggest that CD200 may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms of CD200 have been found. Diseases associated with CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy.
Specifications
Specifications
| Accession Number | O54901 |
| For Use With (Application) | ELISA |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 17470 |
| Molecular Weight (g/mol) | 48.6 kDa |
| Name | CD200 |
| Quantity | 50 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction