missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD177 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8186
This item is not returnable.
View return policy
Description
CD177 Monoclonal specifically detects CD177 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD177 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| CD177, CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1 GP, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1, PRV1NB1 glycoprotein | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 57126 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 1171A | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Q8R2S8 | |
| CD177 | |
| Human embryonic kidney cell line HEK293-derived recombinant mouse CD177 Accession # Q8R2S8 | |
| 100 μg | |
| Primary | |
| Detects mouse CD177 in direct ELISAs. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction