missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD161/NK1.1 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB76141B-100
This item is not returnable.
View return policy
Description
CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD161/NK1.1 | |
| Monoclonal | |
| Biotin | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Klrb1c, Ly55c, Ly-59, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| NK-1+ cells from mouse spleen and bone marrow, Accession # AAH61168 | |
| 100 Tests | |
| Primary | |
| Reconstitution default not known | |
| Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| PK136 | |
| Flow Cytometry 10 uL/10^6 cells | |
| AAH61168 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 17059 | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido