missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse CD155 (PVR) His-tag Fc Chimera Recombinant Protein

Product Code. 17174283
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 17174283

Brand: Invitrogen™ A42623

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Functional Assay, Immunohistochemistry, Western Blot
Formulation PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 52118
Molecular Weight (g/mol) 63 kDa
Name Mouse CD155 (PVR) His-tag Fc Chimera
Quantity 50 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/ μg
Gene Alias 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4
Common Name CD155
Gene Symbol PVR
Biological Activity Measured by its binding ability in a functional ELISA. Immobilized mouse DNAM1 (at 2 μg/mL (100 μL/well) can bind mouse CD155 with a linear range of 6.4-160 ng/mL)
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag, His-tag
Sequence Mouse CD155, amino acids Met1-Arg345 (Accession # NP_081790.1) with a C-terminal His-tagged human IgG1 Fc-tag
Content And Storage -20°Cor to +80°C if preferred
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.