missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD117/c-kit PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
£134.00 - £252.00
Specifications
| Antigen | CD117/c-kit |
|---|---|
| Clone | 180627 |
| Dilution | Flow Cytometry 10 μL/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18708088
|
R&D Systems
FAB1356P-025 |
25 Tests |
£134.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18770148
|
R&D Systems
FAB1356P-100 |
100 Tests |
£252.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Mouse samples. It is validated for Flow Cytometry.Specifications
| CD117/c-kit | |
| Flow Cytometry 10 μL/106 cells | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P05532 | |
| 3815.0 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SCF R, Gln25-Thr519 (Ala207Glu), Accession No. P05532 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
| 180627 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| KIT | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse SCF R in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title