missing translation for 'onlineSavingsMsg'
Learn More

GIBCO™ Mouse (CD-1) S9 Fractions

Product Code. 15451194 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451194 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15451194 Supplier GIBCO™ Supplier No. MSS9PL
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liver S9 fractions prepared from a pool of male CD-1 mice. Total protein concentration: 20mg/mL

Liver S9 fractions prepared from a pool of male CD-1 mice.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Purity Research Grade
Host Species Mouse
Concentration 20 mg/mL
Product Type Mouse S9 Fraction
Content And Storage Store between -60°C and -85°C.
Format Tube
Sub Cellular Localization S9 Fraction
Shipping Condition Dry Ice
Product Line Gibco
Donor Source Pooled
Gender Male
Physical Form Frozen
Tissue Liver
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.