missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR10 Antibody, R&D Systems™

Rat Monoclonal Antibody
£205.00 - £407.00
Specifications
| Antigen | CCR10/GPR2 |
|---|---|
| Clone | 248918R |
| Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
| Applications | CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726978
![]() |
R&D Systems
MAB2815R-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18727958
![]() |
R&D Systems
MAB2815R-100 |
100 μg |
£407.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CCR10/GPR2 | |
| Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| NP_031747 | |
| 2826 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 248918R | |
| CyTOF, Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 | |
| C6 rat glioma cell line transfected with mouse CCR10 Met1-Asn362 Accession # NP_031747 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects mouse CCR10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title