missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5986R-SP
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CCR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession # P51675 | |
| 25 μg | |
| 1230 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 643854R | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| P51675 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse CCR1 in direct ELISAs. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction