missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5986-SP
This item is not returnable.
View return policy
Description
CCR1 Polyclonal specifically detects CCR1 in Mouse samples. It is validated for Western Blot.
Specifications
| CCR1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1230 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAH11092 | |
| CCR1 | |
| E. coli-derived recombinant mouse CCR1 Met1-Ala34, Asp92-Lys107, Phe172-Arg197, Gln265-Leu281 Accession # AAH11092 | |
| 25 μg | |
| Primary | |
| Detects mouse CCR1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) CCR4, rmCCR5, and rmCCR8 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction