missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL9/10/MIP-1 gamma Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF463
This item is not returnable.
View return policy
Description
CCL9/10/MIP-1 gamma Polyclonal specifically detects CCL9/10/MIP-1 gamma in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CCL9/10/MIP-1 gamma | |
| Polyclonal | |
| Biotin | |
| Q3U9T8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 20308 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| Ccl9 | |
| E. coli-derived recombinant mouse CCL9/10/MIP-1 gamma (R&D Systems, Catalog # 463-MG) Gln22-Gln122 Accession # Q3U9T8 | |
| 50 μg | |
| Primary | |
| Detects mouse CCL9/10/MIP-1 gamma in ELISAs and Western blots. In sandwich immunoassays, less than 0.02% cross-reactivity with recombinant viral (rv) MIP-I, rvMIP-II, recombinant human (rh) MIP-1 alpha , rhMIP-1 beta , rhMIP-1 delta , rhMIP-3 alpha , rhMIP-3 beta , recombinant mouse (rm) MIP-1 alpha , rmMIP-1 beta , recombinant rat MIP-3 alpha , and rmMIP-3 beta is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction