missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CCL8 (MCP-2) Recombinant Protein, PeproTech®

Product Code. 17899962
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17899962

Brand: Gibco™ 25014100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MCP-2 is an 8.5 kDa protein containing 74 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z121
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20307
Molecular Weight (g/mol) 8.5 kDa
Name Mouse CCL8 (MCP-2)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1810063B20Rik; AB023418; C-C motif chemokine 8; C-C motif chemokine ligand 8; CCL8; chemokine (C-C motif) ligand 8; HC14; MCP2; MCP-2; MCP-2(6-76); monocyte chemoattractant protein 2; monocyte chemoattractant protein-2; monocyte chemotactic protein 2; SCYA10; SCYA8; small inducible cytokine A8; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); small-inducible cytokine A8
Common Name CCL8 (MCP2)
Gene Symbol CCL8
Biological Activity Determined by its ability to chemoattract human peripheral blood monocytes using a concentration range of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.